Assay Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019635.2 | NP_062609.2 | 4 - 5 | 602 | 123 | |
| GenBank mRNA | AF271361.1 | - | 4 - 5 | 348 | 123 | |
| AY058922.1 | - | 4 - 5 | 488 | 123 | ||
| BC037440.1 | - | 2 - 3 | 358 | 3585312 | 123 | |
| BC049123.1 | - | 4 - 5 | 476 | 6813956 | 123 | |
| U28726.1 | - | 4 - 5 | 481 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk3
Genomic Map