Assay Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013792.2 | NP_001013814.2 | 1 - 2 | 238 | 109 | |
| GenBank mRNA | AK134955.1 | - | 1 - 2 | 73 | 109 | |
| AK150371.1 | - | 1 - 2 | 241 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134955.1 transcripts.
Set Membership: |
|

Otulin
Genomic Map