Assay Details
| Gene Symbol: Hinfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172162.3 | NP_751894.2 | 2 - 3 | 237 | 76 | |
| GenBank mRNA | AK049348.1 | - | 2 - 3 | 201 | 76 | |
| AK049633.1 | - | 2 - 3 | 398 | 76 | ||
| BC029091.1 | - | 1 - 2 | 220 | 2651006 | 76 | |
| BY196765.1 | - | 2 - 3 | 237 | 76 | ||
| CN708951.1 | - | 1 - 2 | 83 | 76 | ||
| CT010239.1 | - | 1 - 2 | 172 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hinfp
Genomic Map