Assay Details
| Gene Symbol: C2cd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027909.2 | NP_082185.2 | 3 - 4 | 969 | 68 | |
| GenBank mRNA | AK004920.1 | - | 3 - 4 | 974 | 68 | |
| AK039220.1 | - | 3 - 4 | 940 | 68 | ||
| AK122237.1 | - | 3 - 4 | 915 | 68 | ||
| AK153804.1 | - | 3 - 4 | 941 | 68 | ||
| BC035271.1 | - | 2 - 3 | 209 | 68 | ||
| BC056612.1 | - | 3 - 4 | 540 | 4952570 | 68 | |
| BC060156.1 | - | 3 - 4 | 833 | 6810220 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

C2cd2l
Genomic Map