Assay Details
| Gene Symbol: Prtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011178.2 | NP_035308.2 | 3 - 4 | 385 | 81 | |
| GenBank mRNA | FM210470.1 | - | 1 - 2 | 199 | 81 | |
| U43525.1 | - | 3 - 4 | 373 | 81 | ||
| U97073.1 | - | 3 - 4 | 385 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the FM210470.1 transcripts.
Set Membership: |
|

Prtn3
Genomic Map