Assay Details
| Gene Symbol: Lgi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144556.2 | NP_653139.2 | 5 - 6 | 545 | 58 | |
| GenBank mRNA | AJ487521.1 | - | 5 - 6 | 546 | 58 | |
| AJ487960.1 | - | 5 - 6 | 545 | 58 | ||
| AK033967.1 | - | 5 - 6 | 363 | 58 | ||
| AK134643.1 | - | 5 - 6 | 1139 | 58 | ||
| AK148793.1 | - | 5 - 6 | 804 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgi4
Genomic Map