Assay Details
| Gene Symbol: Lgi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144556.2 | NP_653139.2 | 7 - 8 | 879 | 72 | |
| GenBank mRNA | AJ487521.1 | - | 7 - 8 | 880 | 72 | |
| AJ487960.1 | - | 7 - 8 | 879 | 72 | ||
| AK033967.1 | - | 7 - 8 | 697 | 72 | ||
| AK134643.1 | - | 7 - 8 | 1473 | 72 | ||
| AK148793.1 | - | 7 - 8 | 1139 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgi4
Genomic Map