Assay Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081127.1 | NP_001074596.1 | 19 - 20 | 3100 | 114 | |
| GenBank mRNA | AK137573.1 | - | 4 - 5 | 552 | 114 | |
| BC158014.1 | - | 19 - 20 | 2933 | 9088151 | 114 | |
| BC172106.1 | - | 19 - 20 | 2916 | 114 | ||
More Information
Set Membership: |
|

Adamts14
Genomic Map