Assay Details
| Gene Symbol: Tvp23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013778.1 | NP_001013800.1 | 2 - 3 | 565 | 61 | |
| GenBank mRNA | BC068110.1 | - | 2 - 3 | 565 | 5696118 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Tvp23a
Genomic Map