| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900016293.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016293.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016293.mm.1 |
TC0900001940.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016291.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016291.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016291.mm.1 |
TC0900001940.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016294.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016294.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016294.mm.1 |
TC0900001940.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016292.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016292.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016292.mm.1 |
TC0900001940.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016295.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016295.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016295.mm.1 |
TC0900001940.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016296.mm.1 |
NM_001113323.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016296.mm.1 |
NM_001113323.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016296.mm.1 |
TC0900001940.mm.1 |
MTA TC |