Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025615.3 | NP_001020786.1 | 3 - 4 | 369 | 75 | |
| NM_026202.4 | NP_080478.2 | 3 - 4 | 369 | 75 | ||
| GenBank mRNA | AJ534985.1 | - | 3 - 4 | 236 | 75 | |
| AK012190.1 | - | 3 - 4 | 263 | 75 | ||
| AK016827.1 | - | 3 - 4 | 344 | 75 | ||
| AK147820.1 | - | 3 - 4 | 320 | 75 | ||
| AK151672.1 | - | 3 - 4 | 331 | 75 | ||
| AK162434.1 | - | 3 - 4 | 346 | 75 | ||
| AK165214.1 | - | 3 - 4 | 274 | 75 | ||
| BC144895.1 | - | 3 - 4 | 348 | 9053443 | 75 | |
| BC145933.1 | - | 3 - 4 | 348 | 40131236 | 75 | |
| BC145935.1 | - | 3 - 4 | 348 | 40131238 | 75 | |
| BY716501.1 | - | 3 - 4 | 344 | 75 | ||
| CJ045424.1 | - | 3 - 4 | 373 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc50
Genomic Map