Assay Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144936.1 | NP_659185.1 | 4 - 5 | 666 | 67 | |
| GenBank mRNA | AK079262.1 | - | 4 - 5 | 653 | 67 | |
| AK144325.1 | - | 4 - 5 | 891 | 67 | ||
| BC018222.1 | - | 4 - 5 | 622 | 4221959 | 67 | |
| BC026654.1 | - | 4 - 5 | 666 | 4988382 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45b
Genomic Map