Assay Details
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161623.1 | NP_001155095.1 | 5 - 6 | 474 | 82 | |
| NM_009862.2 | NP_033992.2 | 7 - 8 | 851 | 82 | ||
| GenBank mRNA | AF098068.1 | - | 6 - 7 | 597 | 82 | |
| AK147013.1 | - | 6 - 7 | 568 | 82 | ||
| AK167850.1 | - | 5 - 6 | 462 | 82 | ||
| BB852042.1 | - | 5 - 6 | 425 | 82 | ||
| BC006856.1 | - | 6 - 7 | 568 | 82 | ||
| BC028635.1 | - | 6 - 7 | 604 | 1262927 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc45
Genomic Map