Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 12 - 13 | 1841 | 85 | |
| NM_001190975.1 | NP_001177904.1 | 11 - 12 | 1577 | 85 | ||
| NM_009465.4 | NP_033491.2 | 13 - 14 | 1868 | 85 | ||
| GenBank mRNA | AK033237.1 | - | 12 - 13 | 1810 | 85 | |
| AK155567.1 | - | 14 - 15 | 1754 | 85 | ||
| AK161294.1 | - | 6 - 7 | 541 | 85 | ||
| AK170897.1 | - | 12 - 13 | 86 | |||
| AK199775.1 | - | 1 - 2 | 48 | 85 | ||
| BC046618.1 | - | 13 - 14 | 1705 | 6494383 | 85 | |
| BC050914.1 | - | 13 - 14 | 1676 | 6418252 | 85 | |
| BC058230.1 | - | 13 - 14 | 1729 | 6313662 | 85 | |
| X63535.1 | - | 13 - 14 | 1617 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Axl
Genomic Map