Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 2 - 3 | 545 | 72 | |
| NM_001190975.1 | NP_001177904.1 | 2 - 3 | 545 | 72 | ||
| NM_009465.4 | NP_033491.2 | 2 - 3 | 545 | 72 | ||
| GenBank mRNA | AK033237.1 | - | 2 - 3 | 514 | 72 | |
| AK155567.1 | - | 3 - 4 | 431 | 72 | ||
| AK170010.1 | - | 2 - 3 | 519 | 72 | ||
| AK170897.1 | - | 3 - 4 | 442 | 72 | ||
| BC046618.1 | - | 2 - 3 | 382 | 6494383 | 72 | |
| BC050914.1 | - | 2 - 3 | 353 | 6418252 | 72 | |
| BC058230.1 | - | 3 - 4 | 433 | 6313662 | 72 | |
| X63535.1 | - | 2 - 3 | 294 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Axl
Genomic Map