Assay Details
| Gene Symbol: Paqr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028748.2 | NP_083024.1 | 5 - 6 | 706 | 93 | |
| GenBank mRNA | AK002481.1 | - | 5 - 6 | 676 | 93 | |
| AK035475.1 | - | 5 - 6 | 718 | 93 | ||
| AK085411.1 | - | 5 - 6 | 707 | 93 | ||
| AK086679.1 | - | 5 - 6 | 720 | 93 | ||
| AY424294.1 | - | 5 - 6 | 676 | 93 | ||
| BC054855.1 | - | 5 - 6 | 732 | 93 | ||
| BC100516.1 | - | 4 - 5 | 587 | 30919009 | 93 | |
More Information
Set Membership: |
|

Paqr5
Genomic Map