Assay Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134109.2 | NP_598870.1 | 7 - 8 | 1714 | 65 | |
| GenBank mRNA | AK036844.1 | - | 7 - 8 | 1640 | 65 | |
| BC010485.1 | - | 7 - 8 | 1654 | 3483341 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ildr1
Genomic Map