Assay Details
| Gene Symbol: Rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145480.1 | NP_663455.1 | 2 - 3 | 193 | 76 | |
| GenBank mRNA | AK146954.1 | - | 2 - 3 | 181 | 76 | |
| AK147030.1 | - | 2 - 3 | 181 | 76 | ||
| AK196924.1 | - | 1 - 2 | 79 | 76 | ||
| BC003335.1 | - | 2 - 3 | 193 | 3257208 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc4
Genomic Map