Assay Details
| Gene Symbol: Rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145480.1 | NP_663455.1 | 7 - 8 | 733 | 99 | |
| GenBank mRNA | AK146954.1 | - | 7 - 8 | 721 | 99 | |
| AK147030.1 | - | 7 - 8 | 721 | 99 | ||
| BC003335.1 | - | 7 - 8 | 733 | 3257208 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rfc4
Genomic Map