Assay Details
| Gene Symbol: Htr1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008310.3 | NP_032336.1 | 1 - 2 | 158 | 105 | |
| GenBank mRNA | AK045364.1 | - | 1 - 2 | 227 | 105 | |
| Z14224.1 | - | 1 - 2 | 158 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045364.1, NM_008310.3, Z14224.1 transcripts.
Set Membership: |
|

Htr1f
Genomic Map