Assay Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145615.4 | NP_663590.3 | 10 - 11 | 1056 | 98 | |
| GenBank mRNA | AK028118.1 | - | 10 - 11 | 1057 | 98 | |
| AK032830.1 | - | 10 - 11 | 1009 | 98 | ||
| AK133525.1 | - | 10 - 11 | 1002 | 98 | ||
| AK167572.1 | - | 10 - 11 | 1029 | 98 | ||
| AK168321.1 | - | 10 - 11 | 1017 | 98 | ||
| AV289509.2 | - | 4 - 5 | 237 | 98 | ||
| BC096645.1 | - | 10 - 11 | 915 | 1245213 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Etfa
Genomic Map