Assay Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145615.4 | NP_663590.3 | 6 - 7 | 737 | 68 | |
| GenBank mRNA | AK028118.1 | - | 6 - 7 | 738 | 68 | |
| AK032830.1 | - | 6 - 7 | 690 | 68 | ||
| AK133525.1 | - | 6 - 7 | 683 | 68 | ||
| AK167572.1 | - | 6 - 7 | 710 | 68 | ||
| AK168321.1 | - | 6 - 7 | 698 | 68 | ||
| BC003432.1 | - | 6 - 7 | 628 | 2646522 | 68 | |
| BC096645.1 | - | 6 - 7 | 596 | 1245213 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Etfa
Genomic Map