Assay Details
| Gene Symbol: Pla1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134102.4 | NP_598863.3 | 2 - 3 | 544 | 123 | |
| GenBank mRNA | AF063498.1 | - | 2 - 3 | 545 | 123 | |
| BC003470.1 | - | 2 - 3 | 287 | 2649848 | 123 | |
| BC030670.1 | - | 2 - 3 | 543 | 1247206 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla1a
Genomic Map