Assay Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009840.3 | NP_033970.3 | 8 - 9 | 1018 | 85 | |
| GenBank mRNA | AK010882.1 | - | 8 - 9 | 1019 | 85 | |
| AK014005.1 | - | 1 - 2 | 84 | 85 | ||
| AK075711.1 | - | 8 - 9 | 999 | 85 | ||
| AK075880.1 | - | 8 - 9 | 1007 | 85 | ||
| AK145661.1 | - | 8 - 9 | 1002 | 85 | ||
| AK145754.1 | - | 8 - 9 | 1001 | 85 | ||
| AK145918.1 | - | 8 - 9 | 1020 | 85 | ||
| AK147038.1 | - | 8 - 9 | 1021 | 85 | ||
| AK159969.1 | - | 8 - 9 | 1010 | 85 | ||
| AK168465.1 | - | 8 - 9 | 991 | 85 | ||
| AK172867.1 | - | 8 - 9 | 989 | 85 | ||
| BC009007.1 | - | 8 - 9 | 976 | 3582639 | 85 | |
| Z37164.1 | - | 8 - 9 | 1022 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct8
Genomic Map