Assay Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080457.3 | NP_536705.3 | 14 - 15 | 9177 | 91 | |
| GenBank mRNA | AF218265.1 | - | 14 - 15 | 2482 | 91 | |
| AF441786.1 | - | 8707 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Muc4
Genomic Map