Assay Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159421.1 | NP_001152893.1 | 3 - 4 | 683 | 110 | |
| NM_001159422.1 | NP_001152894.1 | 4 - 5 | 460 | 110 | ||
| NM_198645.2 | NP_941047.1 | 4 - 5 | 809 | 110 | ||
| GenBank mRNA | AK135450.1 | - | 4 - 5 | 389 | 110 | |
| BB686073.1 | - | 2 - 3 | 124 | 110 | ||
| BC024406.1 | - | 4 - 5 | 373 | 110 | ||
| BC024862.1 | - | 4 - 5 | 784 | 5356370 | 110 | |
| BC053690.1 | - | 4 - 5 | 361 | 110 | ||
| CJ094333.1 | - | 3 - 4 | 264 | 110 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673617 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748128 | BC037188.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55054 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163171 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143129 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29941867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673620 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08563739 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143129 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163171 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08563753 | - | - |
| Whole Mouse Genome G4122A | A_52_P32502 | AK144564.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32502 | AK020822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P404825 | BC024862.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127112 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163171 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143129 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08563753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12599783 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12599792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12599801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12707971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12475463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12707969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12707970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12707968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12656387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12558147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12600682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12537105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12701458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12857641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673687 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10435491 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435492 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435493 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440260_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424465_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10435489 | - | - |
| Mouse Genome 430 2.0 Array | 1424465_at | - | - |
| Mouse Genome 430 2.0 Array | 1424465_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381045_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381045_at | - | - |
| Mouse Expression Set 430 | 1424465_at | - | - |
| Mouse Expression Set 430 | 1424465_PM_at | - | - |
| Murine Genome U74v2 Set | 160297_PM_at | - | - |
| Murine Genome U74v2 Set | 160297_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424465_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424465_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424465_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424465_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440260_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440260_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 460546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 460546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460546 | - | - |
| mouse-6 v1.1 (Illumina) | scl00381045.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00381045.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2949122 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2949122 | - | - |

Ccdc58
Genomic Map