Assay Details
| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011792.5 | NP_035922.4 | 5 - 6 | 1338 | 77 | |
| GenBank mRNA | AF190726.2 | - | 5 - 6 | 1266 | 77 | |
| AF200346.1 | - | 5 - 6 | 839 | 77 | ||
| AK014390.2 | - | 1 - 2 | 137 | 77 | ||
| AK014464.1 | - | 5 - 6 | 1268 | 77 | ||
| AK033112.1 | - | 5 - 6 | 1288 | 77 | ||
| AK041285.1 | - | 5 - 6 | 1288 | 77 | ||
| AK046175.1 | - | 5 - 6 | 1285 | 77 | ||
| AK049626.1 | - | 5 - 6 | 1271 | 77 | ||
| AK079814.1 | - | 4 - 5 | 556 | 77 | ||
| AK080498.1 | - | 5 - 6 | 1290 | 77 | ||
| AK082230.1 | - | 5 - 6 | 1287 | 77 | ||
| AK163583.1 | - | 5 - 6 | 1289 | 77 | ||
| AK173112.1 | - | 3 - 4 | 456 | 77 | ||
| AK197470.1 | - | 207 | 77 | |||
| BC048189.1 | - | 5 - 6 | 1264 | 6831622 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bace1
Genomic Map