Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 3 - 4 | 1218 | 71 | |
| GenBank mRNA | AF330197.1 | - | 3 - 4 | 1214 | 71 | |
| AK036351.1 | - | 2 - 3 | 953 | 71 | ||
| AK048110.1 | - | 2 - 3 | 984 | 71 | ||
| AK137148.1 | - | 1 - 2 | 556 | 71 | ||
| AK169999.1 | - | 2 - 3 | 1328 | 71 | ||
| AK171932.1 | - | 2 - 3 | 1156 | 71 | ||
| BC069835.1 | - | 3 - 4 | 1074 | 30094289 | 71 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map