Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 4 - 5 | 1352 | 92 | |
| GenBank mRNA | AF330197.1 | - | 4 - 5 | 1348 | 92 | |
| AK036351.1 | - | 3 - 4 | 1087 | 92 | ||
| AK048110.1 | - | 3 - 4 | 1118 | 92 | ||
| AK137148.1 | - | 2 - 3 | 690 | 92 | ||
| AK169999.1 | - | 3 - 4 | 1462 | 92 | ||
| AK171932.1 | - | 3 - 4 | 1290 | 92 | ||
| BC069835.1 | - | 4 - 5 | 1208 | 30094289 | 92 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map