Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 7 - 8 | 2028 | 82 | |
| GenBank mRNA | AF330197.1 | - | 7 - 8 | 2024 | 82 | |
| AK036351.1 | - | 6 - 7 | 1763 | 82 | ||
| AK048110.1 | - | 6 - 7 | 1794 | 82 | ||
| AK137148.1 | - | 5 - 6 | 1366 | 82 | ||
| BC011463.1 | - | 1 - 2 | 460 | 82 | ||
| BC069835.1 | - | 7 - 8 | 1884 | 30094289 | 82 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map