Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 8 - 9 | 2284 | 62 | |
| GenBank mRNA | AF330197.1 | - | 8 - 9 | 2280 | 62 | |
| AK035242.1 | - | 2 - 3 | 601 | 62 | ||
| AK036351.1 | - | 7 - 8 | 2019 | 62 | ||
| AK048110.1 | - | 7 - 8 | 2050 | 62 | ||
| AK137148.1 | - | 6 - 7 | 1622 | 62 | ||
| AK169999.1 | - | 7 - 8 | 2390 | 62 | ||
| BC011463.1 | - | 2 - 3 | 716 | 62 | ||
| BC069835.1 | - | 8 - 9 | 2140 | 30094289 | 62 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map