Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 10 - 11 | 2758 | 83 | |
| GenBank mRNA | AF330197.1 | - | 10 - 11 | 2754 | 83 | |
| AK035242.1 | - | 4 - 5 | 1075 | 83 | ||
| AK036351.1 | - | 9 - 10 | 2493 | 83 | ||
| AK048110.1 | - | 9 - 10 | 2524 | 83 | ||
| AK137148.1 | - | 8 - 9 | 2096 | 83 | ||
| AK164502.1 | - | 1 - 2 | 155 | 83 | ||
| AK169999.1 | - | 9 - 10 | 2864 | 83 | ||
| BC026485.1 | - | 1 - 2 | 119 | 83 | ||
| BC069835.1 | - | 10 - 11 | 2614 | 30094289 | 83 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map