Assay Details
| Gene Symbol: Trmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080999.2 | NP_001074468.1 | 4 - 5 | 1211 | 77 | |
| NM_001081000.2 | NP_001074469.1 | 4 - 5 | 1258 | 77 | ||
| NM_001195205.1 | NP_001182134.1 | 4 - 5 | 1211 | 77 | ||
| GenBank mRNA | AK080170.1 | - | 4 - 5 | 1086 | 77 | |
| AK133844.1 | - | 4 - 5 | 1086 | 77 | ||
| AK134368.1 | - | 4 - 5 | 1212 | 77 | ||
| AK146391.1 | - | 4 - 5 | 1151 | 77 | ||
| AK150482.1 | - | 4 - 5 | 1056 | 77 | ||
| AK151311.1 | - | 4 - 5 | 1056 | 77 | ||
| AK151422.1 | - | 4 - 5 | 1082 | 77 | ||
| AK151725.1 | - | 4 - 5 | 1056 | 77 | ||
| AK151778.1 | - | 4 - 5 | 1056 | 77 | ||
| AK152055.1 | - | 4 - 5 | 1056 | 77 | ||
| AK152201.1 | - | 4 - 5 | 1056 | 77 | ||
| AK152363.1 | - | 4 - 5 | 1056 | 77 | ||
| AK152548.1 | - | 4 - 5 | 1056 | 77 | ||
| AK152903.1 | - | 4 - 5 | 1056 | 77 | ||
| AK153043.1 | - | 4 - 5 | 1056 | 77 | ||
| AK153190.1 | - | 4 - 5 | 1082 | 77 | ||
| AK181536.1 | - | 1 - 2 | 52 | 77 | ||
| AK182911.1 | - | 1 - 2 | 113 | 77 | ||
| AK185507.1 | - | 1 - 2 | 113 | 77 | ||
| AK206481.1 | - | 1 - 2 | 113 | 77 | ||
| BC027431.1 | - | 3 - 4 | 1323 | 3156359 | 77 | |
| BC038486.1 | - | 4 - 5 | 1084 | 3155739 | 77 | |
| BC046976.1 | - | 4 - 5 | 1092 | 6337243 | 77 | |
| X56044.1 | - | 3 - 4 | 1655 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trmt2a
Genomic Map