Assay Details
| Gene Symbol: Med15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040683.2 | NP_001035773.1 | 10 - 11 | 1545 | 61 | |
| NM_001285884.1 | NP_001272813.1 | 11 - 12 | 1665 | 61 | ||
| NM_001285886.1 | NP_001272815.1 | 10 - 11 | 1357 | 61 | ||
| NM_033609.3 | NP_291087.2 | 11 - 12 | 1665 | 61 | ||
| GenBank mRNA | AF328770.1 | - | 11 - 12 | 1541 | 61 | |
| AK131186.1 | - | 10 - 11 | 1486 | 61 | ||
| AK142224.1 | - | 11 - 12 | 1632 | 61 | ||
| AK166522.1 | - | 10 - 11 | 1357 | 61 | ||
| AK169904.1 | - | 11 - 12 | 1666 | 61 | ||
| BC003221.1 | - | 1 - 2 | 104 | 61 | ||
| BC049891.1 | - | 6 - 7 | 1129 | 61 | ||
| BC054779.1 | - | 10 - 11 | 1495 | 6812541 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med15
Genomic Map