Assay Details
| Gene Symbol: Med15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040683.2 | NP_001035773.1 | 15 - 16 | 2137 | 89 | |
| NM_001285884.1 | NP_001272813.1 | 16 - 17 | 2257 | 89 | ||
| NM_001285886.1 | NP_001272815.1 | 15 - 16 | 1949 | 89 | ||
| NM_033609.3 | NP_291087.2 | 16 - 17 | 2257 | 89 | ||
| GenBank mRNA | AF328770.1 | - | 16 - 17 | 2134 | 89 | |
| AK131186.1 | - | 15 - 16 | 2078 | 89 | ||
| AK142224.1 | - | 16 - 17 | 2224 | 89 | ||
| AK166522.1 | - | 15 - 16 | 1949 | 89 | ||
| AK169904.1 | - | 16 - 17 | 2258 | 89 | ||
| BC003221.1 | - | 6 - 7 | 696 | 89 | ||
| BC049891.1 | - | 11 - 12 | 1721 | 89 | ||
| BC054779.1 | - | 15 - 16 | 2087 | 6812541 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med15
Genomic Map