Assay Details
| Gene Symbol: Smpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164609.1 | NP_001158081.1 | 17 - 18 | 1955 | 111 | |
| NM_001164610.1 | NP_001158082.1 | 16 - 17 | 1916 | 111 | ||
| NM_001164611.1 | NP_001158083.1 | 16 - 17 | 1913 | 111 | ||
| NM_029945.3 | NP_084221.2 | 17 - 18 | 2003 | 111 | ||
| GenBank mRNA | AK019466.2 | - | 17 - 18 | 2004 | 111 | |
| AK031029.1 | - | 16 - 17 | 1907 | 111 | ||
| AK031685.1 | - | 16 - 17 | 1915 | 111 | ||
| AK035892.1 | - | 16 - 17 | 1908 | 111 | ||
| AK036055.1 | - | 16 - 17 | 1908 | 111 | ||
| AK036380.1 | - | 16 - 17 | 1912 | 111 | ||
| AK129354.1 | - | 17 - 18 | 1980 | 111 | ||
| AK164234.1 | - | 17 - 18 | 1947 | 111 | ||
| AK164456.1 | - | 14 - 15 | 2381 | 111 | ||
| BC026767.1 | - | 16 - 17 | 1889 | 5137008 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd4
Genomic Map