Assay Details
| Gene Symbol: Smpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164609.1 | NP_001158081.1 | 18 - 19 | 2089 | 103 | |
| NM_001164610.1 | NP_001158082.1 | 17 - 18 | 2050 | 103 | ||
| NM_001164611.1 | NP_001158083.1 | 17 - 18 | 2047 | 103 | ||
| NM_029945.3 | NP_084221.2 | 18 - 19 | 2137 | 103 | ||
| GenBank mRNA | AK019466.2 | - | 18 - 19 | 2138 | 103 | |
| AK031029.1 | - | 17 - 18 | 2041 | 103 | ||
| AK031685.1 | - | 17 - 18 | 2049 | 103 | ||
| AK035892.1 | - | 17 - 18 | 2042 | 103 | ||
| AK036055.1 | - | 17 - 18 | 2042 | 103 | ||
| AK036380.1 | - | 17 - 18 | 2046 | 103 | ||
| AK129354.1 | - | 18 - 19 | 2114 | 103 | ||
| AK164234.1 | - | 18 - 19 | 2081 | 103 | ||
| AK164456.1 | - | 15 - 16 | 2515 | 103 | ||
| BC026767.1 | - | 17 - 18 | 2023 | 5137008 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd4
Genomic Map