Assay Details
| Gene Symbol: Rpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024212.4 | NP_077174.1 | 6 - 7 | 717 | 128 | |
| GenBank mRNA | AK007625.1 | - | 6 - 7 | 730 | 127 | |
| AK008098.1 | - | 6 - 7 | 730 | 128 | ||
| AK011068.1 | - | 6 - 7 | 726 | 128 | ||
| AK012432.1 | - | 6 - 7 | 730 | 128 | ||
| AK050639.1 | - | 6 - 7 | 731 | 128 | ||
| AK088279.1 | - | 6 - 7 | 729 | 128 | ||
| AK135186.1 | - | 6 - 7 | 732 | 128 | ||
| AK151950.1 | - | 6 - 7 | 727 | 128 | ||
| AK165962.1 | - | 6 - 7 | 731 | 128 | ||
| AK166748.1 | - | 6 - 7 | 730 | 128 | ||
| AK166957.1 | - | 6 - 7 | 734 | 128 | ||
| AK167054.1 | - | 6 - 7 | 732 | 128 | ||
| BC003459.1 | - | 6 - 7 | 714 | 2648518 | 128 | |
| BC094036.1 | - | 6 - 7 | 712 | 5011554 | 128 | |
| BC132321.1 | - | 6 - 7 | 717 | 40130598 | 128 | |
| BC132323.1 | - | 6 - 7 | 717 | 40130600 | 128 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl4
Genomic Map