Assay Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016787.4 | NP_058067.2 | 9 - 10 | 1165 | 143 | |
| GenBank mRNA | AF035207.1 | - | 9 - 10 | 1017 | 143 | |
| AF400107.1 | - | 8 - 9 | 905 | 143 | ||
| AK054288.1 | - | 9 - 10 | 1131 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bnip2
Genomic Map