Assay Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022986.4 | NP_075362.3 | 2 - 3 | 598 | 157 | |
| GenBank mRNA | AF093135.1 | - | 2 - 3 | 381 | 157 | |
| AJ242722.1 | - | 2 - 3 | 157 | |||
| AK013036.2 | - | 2 - 3 | 403 | 157 | ||
| AK020170.1 | - | 2 - 3 | 601 | 157 | ||
| AK020448.1 | - | 2 - 3 | 373 | 157 | ||
| AK045436.1 | - | 2 - 3 | 394 | 157 | ||
| AK076594.1 | - | 2 - 3 | 406 | 157 | ||
| BC046301.1 | - | 2 - 3 | 392 | 157 | ||
| BC061099.1 | - | 2 - 3 | 385 | 6742251 | 157 | |
More Information
Set Membership: |
|

Irak1bp1
Genomic Map