Assay Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039522.1 | NP_001034611.1 | 1 - 2 | 119 | 78 | |
| GenBank mRNA | BC082540.1 | - | 1 - 2 | 119 | 78 | |
| BC083358.1 | - | 1 - 2 | 107 | 30625386 | 78 | |
| CB248994.1 | - | 1 - 2 | 119 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Leo1
Genomic Map