Assay Details
| Gene Symbol: Leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039522.1 | NP_001034611.1 | 3 - 4 | 981 | 98 | |
| GenBank mRNA | BC083358.1 | - | 3 - 4 | 969 | 30625386 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Leo1
Genomic Map