Assay Details
| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133718.4 | NP_598479.1 | 3 - 4 | 665 | 111 | |
| GenBank mRNA | AK008408.1 | - | 3 - 4 | 551 | 111 | |
| AK077065.1 | - | 3 - 4 | 668 | 111 | ||
| AK150071.1 | - | 3 - 4 | 568 | 111 | ||
| AK159364.1 | - | 3 - 4 | 592 | 111 | ||
| AK161096.1 | - | 3 - 4 | 547 | 111 | ||
| AK170171.1 | - | 3 - 4 | 568 | 111 | ||
| BC018367.1 | - | 3 - 4 | 517 | 2648510 | 111 | |
| BC018491.1 | - | 3 - 4 | 545 | 2651602 | 111 | |
| BC026136.1 | - | 2 - 3 | 141 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem30a
Genomic Map