Assay Details
| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010304.3 | NP_034434.1 | 2 - 3 | 606 | 94 | |
| GenBank mRNA | BC005439.1 | - | 2 - 3 | 606 | 3592300 | 94 |
| BC011098.1 | - | 2 - 3 | 356 | 3967497 | 94 | |
| M80632.1 | - | 2 - 3 | 377 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gna15
Genomic Map