Assay Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030481.1 | - | 1 - 2 | 61 | 66 | |
| AK171197.1 | - | 49 | 66 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030481.1, AK171197.1 transcripts.
Set Membership: |
|

Mob3a
Genomic Map