Assay Details
| Gene Symbol: Fzr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019757.1 | NP_062731.1 | 12 - 13 | 1496 | 74 | |
| GenBank mRNA | AF083809.1 | - | 12 - 13 | 1496 | 74 | |
| AK051993.1 | - | 12 - 13 | 1419 | 74 | ||
| AK081476.1 | - | 12 - 13 | 1552 | 74 | ||
| AK083686.1 | - | 1566 | 74 | |||
| AK154822.1 | - | 12 - 13 | 1580 | 74 | ||
| AK155370.1 | - | 12 - 13 | 1578 | 74 | ||
| AK163951.1 | - | 12 - 13 | 1555 | 74 | ||
| AK172679.1 | - | 9 - 10 | 2516 | 74 | ||
| AK185038.1 | - | 2 - 3 | 217 | 74 | ||
| BC006616.1 | - | 12 - 13 | 1577 | 3967009 | 74 | |
| FJ786633.2 | - | 7 - 8 | 730 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fzr1
Genomic Map