Assay Details
| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010295.2 | NP_034425.1 | 2 - 3 | 540 | 106 | |
| GenBank mRNA | AK160876.1 | - | 2 - 3 | 300 | 106 | |
| AK169676.1 | - | 2 - 3 | 513 | 106 | ||
| BC019374.1 | - | 2 - 3 | 297 | 4195425 | 106 | |
| U85414.1 | - | 2 - 3 | 265 | 106 | ||
| U85498.1 | - | 2 - 3 | 265 | 106 | ||
More Information
Set Membership: |
|

Gclc
Genomic Map