Assay Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008103.3 | NP_032129.2 | 3 - 4 | 513 | 86 | |
| GenBank mRNA | AK142574.1 | - | 3 - 4 | 499 | 86 | |
| BC066866.1 | - | 3 - 4 | 513 | 30432193 | 86 | |
| D88612.1 | - | 3 - 4 | 548 | 86 | ||
| U59876.1 | - | 3 - 4 | 529 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcm1
Genomic Map