Assay Details
| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011827.3 | NP_035957.2 | 1 - 2 | 147 | 56 | |
| GenBank mRNA | AF072846.1 | - | 1 - 2 | 39 | 56 | |
| AF172930.1 | - | 1 - 2 | 147 | 56 | ||
| AK008005.1 | - | 1 - 2 | 146 | 56 | ||
| BC145545.1 | - | 1 - 2 | 138 | 9054109 | 56 | |
| BC145824.1 | - | 1 - 2 | 138 | 40131127 | 56 | |
| BC145826.1 | - | 1 - 2 | 138 | 40131129 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031432 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984886 | BC069220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984886 | NM_011827.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984886 | AF172930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984886 | AK008005.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78262 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131230 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984886 | BC069220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984886 | NM_011827.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984886 | AF172930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984886 | AK008005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P170188 | BC145824.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P170188 | BC145826.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P170188 | BC069220.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P170188 | NM_011827.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P170188 | AF072846.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131230 | - | - |
| Whole Mouse Genome G4122A | A_52_P170188 | BC145824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170188 | BC145826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170188 | BC069220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170188 | NM_011827.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170188 | AF072846.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116120 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123931 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06118270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06195727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06312801 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06312796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06169271 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06118272 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06053381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06053381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06312795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25031408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561925 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419119_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561920 | - | - |
| Mouse Genome 430 2.0 Array | 1419119_at | - | - |
| Mouse Genome 430 2.0 Array | 1419119_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23900_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23900_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700024453.mm.1 | NM_011827.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024453.mm.1 | NM_011827.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024453.mm.1 | TC0700002758.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024454.mm.1 | NM_011827.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024454.mm.1 | NM_011827.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024454.mm.1 | TC0700002758.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419119_at | - | - |
| Mouse Expression Set 430 | 1419119_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419119_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419119_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419119_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419119_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 849375 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104590020 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104590020 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2350603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218132 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220418 | - | - |
| mouse-6 v1.1 (Illumina) | scl31527.4.1_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0023900.2_111-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350603 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104590020 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0023900.2_111-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104590020 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220418 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220418 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0023900.2_111-S | - | - |

Hcst
Genomic Map