Assay Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138676.2 | NP_619617.2 | 15 - 16 | 1639 | 99 | |
| GenBank mRNA | AF246218.1 | - | 15 - 16 | 1645 | 99 | |
| AK047434.1 | - | 13 - 14 | 1770 | 99 | ||
| AK047826.1 | - | 3 - 4 | 438 | 99 | ||
| AK154779.1 | - | 15 - 16 | 1627 | 99 | ||
| AK160641.1 | - | 14 - 15 | 1711 | 99 | ||
| AK187576.1 | - | 2 - 3 | 170 | 99 | ||
| BC061477.1 | - | 15 - 16 | 1592 | 6336438 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shkbp1
Genomic Map